Order Entry
India
ContactUsLinkComponent
Water for HPLC, Rankem®
Water for HPLC, Rankem®
Catalog # RANKW0020
Supplier:  RANKEM
CAS Number:  
Water for HPLC, Rankem®
Catalog # RANKW0020
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Amber coloured PET bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Guarantee Analysis:
    .
  • Non volatile matter:
    Max. 0.0003 %
  • UV Absorbance:
    .
  • Wavelength:
    .
  • 210 nm:
    Max. 0.004
  • 254 nm:
    Max. 0.002

Specification Test Results

About this item

 :
 :18.02 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332