
Sodium hydroxide solution N/10, Rankem®
Catalog # RANKS0770
Supplier: RANKEM
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:40 g/mol
Storage Temperature:Ambient
UN:1824
ADR:8,III
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548