

2-Propanol ≥99.80%, Electronic Grade, Rankem®
Catalog # RANKP2765
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber coloured glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):≥ 99.80%
- Weight Per ml (at 20 °C):0.785 - 0.787 g
- Refractive Index (at 20 °C):1.377 - 1.378
- Distillation Range:81.0 - 83.0 °C
- Maximum Limits Of Impurities:.
- Non volatile matter:≤ 0.0005%
- Acidity (As C2H5COOH):≤ 0.001%
- Alkalinity (As Nh3):≤ 0.0005%
- Aldehydes And Ketones:<= 0.005%
- Calcium (Ca):≤ 0.02 ppm
- Chloride (Cl):<= 0.0001%
- Copper(Cu):≤ 0.01 ppm
- Iron (Fe):≤ 0.01 ppm
- Lead ( Pb):≤ 0.02 ppm
- Magnesium (Mg):≤ 0.01 ppm
- Nickel (Ni):≤ 0.01 ppm
- Substances Reducing Permanganate:<= 0.0005%
- Sulphate (SO4):<= 0.0001%
- Zinc (Zn):≤ 0.01 ppm
- Water:≤ 0.05 %
- Aluminium(Al):≤ 0.05 ppm
- Barium (Ba):≤ 0.01 ppm
- Boron(B):<= 0.0001%
- Cadmium (Cd):≤ 0.005 ppm
- Chromium (Cr):≤ 0.50 ppm
- Cobalt (Co):≤ 0.005 ppm
- Manganese (Mn):≤ 0.005 ppm
- Potassium(K):≤ 0.05 ppm
- Sodium (Na):≤ 0.20 ppm
- Strontium (Sr):<= 0.0001%
- Carbonyl Compounds:<=0.001%
- Methanol:≤ 0.10 %
Specification Test Results
About this item
:
:60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:12 °C
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674