Order Entry
India
ContactUsLinkComponent
Nitric acid 69,00-72,00%, Laboratory Reagent, Rankem®
Oxidizing
Corrosive
Toxic
Nitric acid 69,00-72,00%, Laboratory Reagent, Rankem®
Supplier:  RANKEM
Certificates
Oxidizing
Corrosive
Toxic
Nitric acid 69,00-72,00%, Laboratory Reagent, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Assay:
    69.00 - 72.00%
  • Specific Gravity at 25 ° C:
    1.400 - 1.420
  • Residue On Ignition (As SO₄²⁻):
    ≤ 0.01 %
  • Chloride (Cl):
    ≤ 0.001%
  • Sulphate (SO₄²⁻):
    ≤ 0.01%
  • Arsenic(As):
    ≤ 0.0001%
  • Heavy Metal (As Pb):
    ≤ 0.0005%
  • Iron (Fe):
    ≤ 0.005%

Specification Test Results

Related Information

Frequently Bought Together