Order Entry
India
ContactUsLinkComponent
Nitric acid 69,00-72,00%, Analytical reagent (AR), Rankem®
Oxidizing
Corrosive
Toxic
Nitric acid 69,00-72,00%, Analytical reagent (AR), Rankem®
Supplier:  RANKEM
Certificates
Oxidizing
Corrosive
Toxic
Nitric acid 69,00-72,00%, Analytical reagent (AR), Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Assay:
    69.00 --> 72.00%
  • Specific Gravity at 25 ° C:
    1.400 --> 1.420
  • Residue On Ignition (As SO₄²⁻):
    Max. 0.0010 %
  • Chloride (Cl):
    ≤ 0.00005%
  • Sulphate (SO₄²⁻):
    ≤ 0.0002%
  • Arsenic (As):
    ≤ 0.000001%
  • Heavy Metal (As Pb):
    ≤ 0.00002%
  • Iron (Fe):
    ≤ 0.00002%
  • Phosphate (PO₄³⁻):
    ≤ 0.0001%
  • Silicate (SiO₂):
    ≤ 0.00005%
  • Manganese (Mn):
    Max. 0.00004%
  • Copper (Cu):
    Max. 0.00001%

Specification Test Results

Frequently Bought Together