Ammonium sulphate ≥99.00%, Laboratory Reagent, Rankem®
: RANKEM
:7783-20-2
:
:(NH₄)₂SO₄
:MFCD00003391
:132.14 g/mol
- Assay:Min. 99.00%
- pH (5% Aqueous Solution):5.00 - 6.00
- Sulphated Ash:Max. 0.30000%
- Chloride (Cl):≤0.005%
- Iron (Fe):≤0.002%
- Heavy Metal (As Pb):≤0.002%
Frequently Bought Together