Ammonium chloride ≥99.00%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Specification Test Results
- Clarity Solution of 5 g in 50 ml of Water is Clear and Colourless:Passes Test
- Assay:Min. 99.00%
- Residue On Ignition:Max. 0.10000%
- Phosphate (PO₄):≤0.002%
- Sulphate (SO₄):≤0.03%
- Iron (Fe):Max. 0.00050%
- Heavy Metals (As Pb):Max. 0.0005000
Specification Test Results
Related Information
Frequently Bought Together