Order Entry
India
ContactUsLinkComponent
J.T.Baker®, Hematology Controls, 8 Parameter Control
J.T.Baker®, Hematology Controls, 8 Parameter Control
Catalog # 3747.
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Hematology Controls, 8 Parameter Control
Catalog # 3747.
Supplier:  Avantor
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    8 Parameter control, 4×normal, with screw cap, 4×2,5 ml
  • Seq:
    08
Specifications
About this item
In vitro diagnostic hematology control product, intended for professional use only.
Caution: Parameter Controls are to be stored upright at 2-8°C when not in use. Stored at this temperature, the Controls are guaranteed stable until the expiry date.