

Chloroform ≥99.4% for HPLC, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
UN:1888
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Colour (APHA):≤10
- Assay(GC):99.00 - 99.40%
- Alcohol (As Stabiliser):0.600 - 1.000%
- Weight Per ml (at 20 °C):1.474 - 1.478 g
- Identity (IR):Conforms
- Residue After Evaporation:Max. 0.00050%
- Titrable Acid:Max. 0.00030 meq/g
- Titrable Base:Max. 0.00030 meq/g
- Water ( By KF):Max. 0.050%
- Wavelength, 250 nm:Min. 35.00%
- Wavelength, 260 nm:Min. 80.00%
- Wavelength, From 275 nm:Min. 98.00%
Specification Test Results
Related Information
Frequently Bought Together