Rankem®, Volumetric Flasks, Class A, Glass, Clear, Interchangeable Stopper
: RANKEM
Volumetric flask with interchangeable stopper with accuracy as per Class A.
- Borosilicate glass
- Volumetric flasks 1 and 2 ml are of test tube shape
Frequently Bought Together