Rankem® RanDisc™, Syringe Filters, PVDF Membrane
Supplier: RANKEM
Certificates
About this item
These PVDF membrane filters are ideal choice for biological sample solutions filtration for HPLC analysis. They can also be used to clean aqueous HPLC samples.
- Very low protein binding capacity
- Broad chemical compatibility
Frequently Bought Together