Sulphuric acid 10.0 N, Biotech Reagent, J.T.Baker®
Catalog # 0331-18
Supplier: Avantor
CAS Number:
Specifications
- Pk:18 L
- Seq:5267
- Environmentally Preferable:
Specifications
About this item
:
:98.08 g/mol
UN:2796
:GHS05
:GHS07
:GHS08
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:330
Melting Pt:10.38
Density:1.84
ADR:8 II