Order Entry
India
ContactUsLinkComponent
Rankem®, Flasks, Round Bottom, Glass, Clear, Standard Gound Joints, Two Necks
Rankem®, Flasks, Round Bottom, Glass, Clear, Standard Gound Joints, Two Necks
Catalog # RANKFBRB3N-0500-2P
Supplier:  RANKEM
Rankem®, Flasks, Round Bottom, Glass, Clear, Standard Gound Joints, Two Necks
Catalog # RANKFBRB3N-0500-2P
Supplier:  RANKEM
Specifications
  • Capacity:
    500 ml
  • Seq:
    03
  • Pk:
    4
  • Side necks:
    19/26 NS
  • Centre neck:
    24/29 NS
Specifications
About this item
Borosilicate glass, clear, with one centre neck and two parallel side neck.
  • Side neck with an interchangeable joint