Order Entry
India
ContactUsLinkComponent
 
 
 
Hydrogen peroxide solution 3% (by KMnO₄ titration), AR®, Macron Fine Chemicals™
 
undefined
 
 
Hydrogen peroxide solution 3% (by KMnO₄ titration), AR®, Macron Fine Chemicals™
 

 

  • Assay (H₂O₂) (by KMnO₄ titrn):
    3.10 - 3.30 %
  • Arsenic (As):
    ≤0.00005 %
  • Barium (Ba):
    Passes Test
  • Chloride (Cl):
    ≤0.001 %
  • Free Acid (as H₂SO₄):
    ≤0.010 %
  • Heavy Metals (as Pb):
    ≤0.0005 %
  • Iron (Fe):
    ≤0.0005 %
  • Nitrogen Compounds (as N):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤0.003 %
  • Residue after Evaporation:
    ≤0.020 %
  • Sulfate (SO₄):
    ≤0.010 %

 

 

Frequently Bought Together