Order Entry
India
ContactUsLinkComponent
Dichloromethane ≥99.9% (by GC, corrected for water content), ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Health
Irritant
Dichloromethane ≥99.9% (by GC, corrected for water content), ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH077-10
CAS Number:  
undefined
Health
Irritant
Dichloromethane ≥99.9% (by GC, corrected for water content), ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH077-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, PVC-coated
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • ACS - Assay (CH₂Cl₂) (by GC, corrected for water):
    ≥99.9 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.0003 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Free Halogens:
    Passes Test
  • ACS - Water (H₂O)(by coulometry):
    ≤0.01 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm:
    ≤1.0
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.12
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 - 400 nm:
    ≤0.01
  • Liquid Chromatography Suitability Absorbance:
    Passes Test

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39,8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1,322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881