Ethanol ≥99.9% (V), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
ADR:3,II
:Management of Change (MOC) category = N
UN:1170
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- ACS - Assay (by Volume) :≥99.9%
- ACS - Acetone, 2-Propanol :Passes Test
- ACS - Colour (APHA):≤10
- ACS - Methanol (CH₃OH):≤0.1%
- ACS - Residue after Evaporation:≤0.001%
- ACS - Solubility in Water:Passes Test
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Substances Reducing KMnO₄:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Titrable Base (meq/g):≤0.0002
- ACS - Water (H₂O):≤0.2%
- Trace Impurities - Aluminum (Al):≤0.5 ppm
- Trace Impurities - Barium (Ba):≤0.1 ppm
- Trace Impurities - Boron (B):≤0.02 ppm
- Trace Impurities - Cadmium (Cd):≤0.05 ppm
- Trace Impurities - Calcium (Ca):≤0,5 ppm
- Trace Impurities - Chromium (Cr):≤0.02 ppm
- Trace Impurities - Cobalt (Co:≤0.02 ppm
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Iron (Fe):≤0.10 ppm
- Trace Impurities - Lead (Pb):≤0.10 ppm
- Trace Impurities - Magnesium (Mg):≤0.10 ppm
- Trace Impurities - Manganese (Mn):≤0.02 ppm
- Trace Impurities - Nickel (Ni):≤0.02 ppm
- Trace Impurities - Tin (Sn):≤0.1 ppm
- Trace Impurities - Zinc (Zn):≤0.10 ppm
Specification Test Results
Frequently Bought Together