

Dichloromethane ≥99.8% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
: Avantor
:75-09-2
:
:CH₂Cl₂
:MFCD00000881
:84.93 g/mol
:39,..40 °C
:1,32 g/cm³
:1593
:6.1,III
:GHS07
:GHS08
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water):≥99.8 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Chloride (Cl):≤10 ppm
- Water (by KF, coulometric):≤0.02 %
spec_BAKR9266-7K spec_BAKR9266-95 spec_BAKR9266-9D spec_BAKR9266-9T spec_BAKR9266-A4
Product specification
More
Frequently Bought Together