Florisil® 60-100 mesh, Macron Fine Chemicals™
Catalog # MACR6875-04
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE bottle, Mall Polystormor®
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- On U.S. No. 60 Sieve:≤10 %
- Thru U.S. No. 100 Sieve:≤10 %
Specification Test Results
About this item
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
:
:100.38 g/mol
Density:2,51 g/cm³
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249