Total base number titration solvent mixture ASTM D4739, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
:
Density:1,030...1.040 g/cm³ (25 °C)
UN:1992
ADR:3 (6.1),II
:GHS02
:GHS06
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Density (g/mL) at 25°C:1.030 - 1.040
- Water (v/v):0.9 - 1.1 %
- Assay (by GLC) - Isopropanol (v/v):32 - 34 %
- Assay (by GLC) - Toluene (v/v):32 - 34 %
- Assay (by GLC) - Chloroform (v/v):32 - 34 %
Specification Test Results
Frequently Bought Together