J.T.Baker® BAKERBOND Speedisk®, Oil and Grease Standards Kit
Catalog # 8030-00
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 KIT
- Description:Speedisk® oil and grease standards kit
Specifications
About this item
Mixture (51 ml/bottle) of 2 mg/ml stearic acid and 2 mg/ml n-hexadecane in ULTRA RESIANALYZED acetone.
Packaging: Bottles have a rubber stopper and are foil sealed.