
Potassium permanganate 1.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.995 - 1.005
- Normality:0.995 - 1.005
Specification Test Results
Frequently Bought Together