


Chlorobenzene ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:108-90-7
:
Formula:C₆H₅Cl
MDL Number:MFCD00000530
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–45 °C
Density:1,11 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
REACH:01-2119432722-45
:GHS02
:GHS07
:GHS09
Specification Test Results
- ACS - Assay (C₆H₅Cl) (by GC, corrected for water):≥99.5 %
- ACS - Color (APHA):≤30
- ACS - Residue after Evaporation:≤0.02 %
- ACS - Titrable Acid (meq/g):≤0.004
Specification Test Results
Frequently Bought Together