Order Entry
India
ContactUsLinkComponent
FineCare FIA Meter Plus, Fluoroscence Immunochromatographic Immunoassay system
FineCare FIA Meter Plus, Fluoroscence Immunochromatographic Immunoassay system
Catalog # FINCFS-113
Supplier:  FINECARE
FineCare FIA Meter Plus, Fluoroscence Immunochromatographic Immunoassay system
Catalog # FINCFS-113
Supplier:  FINECARE

Specifications

  • Description:
    Fluoroscence Immunoassay test system FINCFS-113-FINECARE PLUS
  • Pk:
    1

Specifications

About this item

Finecare™ FIA Meter Plus is a fluorescence immunochromatographic analysing system which can help diagnose conditions such as infection, diabetes, cardiovascular diseases, renal injury and cancers, etc.
The Finecare™ platform provides reliable and quantitative results of various kinds of analytes in human blood or urine within several minutes.

  • NGSP, CE, IFCC certified.
  • 8” LCD touch screen display
  • LIS connectivity
  • 30000 test results storage
  • Instant results 3-15 mins. Testing time
  • Self-calibration with High accuracy
  • Diverse application sites: Laboratory, ER, Cardiology, ICU, Respiratory, Pediatrics etc.
  • 25+ parameters 9. Internal Temp Control 10. Built in Thermal Printer 11.


Other features
1. Rapid
2. Accurate
3. Convenient
4. Reliable
5. Expansive
6. Unique Quick mode
7. Designed sampler
8. Reliable accuracy
9. Instant Results
10. Room temparature reagent kit storage
11. Built-In Thermal Printer
12. 8 inch LED Screen
13. LIS connection
14. ID Chip Port
15. Test Channel

Certifications: CE Certified.