- Description:Chemiluminescence Immunoassay (CLIA) System CL-900i
- Electrical:Input Power: 500 VA, Power Supply: 100-240V ~50-60Hz
- W×D×H:860×740×560 mm
- Sample volume:10~200 µl
- Measurement principle:Enhanced ALP-AMPPD method
- Weight:132 kg
- Pk:1
CL-900i, is one of the World's smallest, fully automated CLIA analyzers performs up to 180T/Hr. This is the right size for any small to mid workload labs.
- 180 Tests/hr
- 15 Positions for Reagents
- 50 Samples positions
- Ready to Use Wash Buffers
- It is Small in Size: The World’s Smallest fully-auto CLIA analyzer performs up to 180T/Hr.
The instrument is loaded with unique features & user friendliness in abundance such as
a. Easy access to substrate handling Two sets of substrate, non-stop to replace during test
b. Random access for cuvette loading without pause or stop 88*2 cuvettes in one batch. Cuvettes & waste bag status real-time indicating no cuvette jam
c. 4-phase magnetic separation. Consistent performance reliability with enhanced PMT detection system
Non-touch vortexer mixing for reaction solution without any contamination
d. Each reagent kit contain four chambers; Unique Fin design guarantee mixing effect e. Easy for reagent loading & replacing Liquid reagents ready-to-use
2°C~8°C refrigerating. On board stability up to 56 days.
It is Simple:
a. Free of maintenance
b. One key start
c. Easy access operating area locates outside the cover
d. Consumable status real-time indicating; Random access for cuvette loading without pause or stop 88*2 cuvettes in one batch
a. High Pressure Interior Washing Wiper washing for exterior
b. Minimizes Carry-over, less than 0.3 ppm
c. Stainless Probe
d. Enhanced Washing for sensitive assay
Superior Detection efficiency:
a. The system applies new generation ALP-AMPPD system, with Micron super paramagnetic particles (SPMP) platform, alkaline phosphatase labeled reagents and AMPPD substrate. With enhancing reagents, the signal will be greatly increased and background will be largely decreased.
b. Conventional chemiluminescence combines two liquids together to get a reaction (flash of light), which appears quickly and disappears quickly leaving very little time to detect the signal
c. Enzyme-amplified chemiluminescence reaction produces a glow light that lasts longer allowing the system to take multiple readings and so can be sure of accuracy of result
Mindray CL-900i is a Fully Automatic Chemiluminescence Immunoassay (CLIA) System is powerful system with a throughput of 180 Tests/Hr. The Measurement Principle is based on Micron super paramagnetic particles (SPMP) platform with alkaline phosphatase (ALP) labeled reagents and AMPPD substrate.
Further, When there is single separation, the residual of labeled antibody would lead to false positive results; but we employ up to 4-phase wash separation steps, which would lower the possibility to the lowest level.