- Throughput:Up to 120 tests/hour
- Description:Chemiluminescence Immunoassay (CLIA) System CL-1000i
- Electrical:Input Power: 500 VA, Power Supply: 100-240V ~50-60Hz
- W×D×H:1400×760×600 mm
- Sample volume:10 µl ~200 µl, with increment of 1 µl
- Measurement principle:Enhanced ALP-AMPPD measurement method
- Weight:250 kg
- Pk:1
CL-1000i, is a robust and simple to use chemiluminescence analyzer with a throughput of up to 120 tests per hour. In a compact benchtop package, it has a large onboard capacity of 25 reagents and 60 samples, and supports non-stop refill of samples, reagents and consumables during testing. By integrating the reliable CLIA technology with latest electrical and software improvement, CL-1000i enables easy operation and brings confident diagnosis faster than ever.
- 120 Tests/hr
- 25 Positions for Reagents
- 60 Samples positions
- Ready to Use Wash Buffers
It is a Robust and fully capable, benchtop analyzer offering high performance: Fully-auto CLIA analyzer performs up to 120T/Hr.
It is Simple:
a. Free of maintenance
b. One key start
c. Easy access operating area locates outside the cover
d. Consumable status real-time indicating; Random access for cuvette loading without pause or stop 88*2 cuvettes in one batch
a.High Pressure Interior Washing Wiper washing for exterior
b. Minimizes Carry-over, less than 0.3 ppm
c. Stainless Probe
d. Enhanced Washing for sensitive assay
Superior Detection efficiency:
a. The system applies new generation ALP-AMPPD system, with Micron super paramagnetic particles (SPMP) platform, alkaline phosphatase labeled reagents and AMPPD substrate. With enhancing reagents, the signal will be greatly increased and background will be largely decreased.
b. Conventional chemiluminescence combines two liquids together to get a reaction (flash of light), which appears quickly and disappears quickly leaving very little time to detect the signal
c. Enzyme-amplified chemiluminescence reaction produces a glow light that lasts longer allowing the system to take multiple readings and so can be sure of accuracy of result
Mindray CL-1000i is a Fully Automatic Chemiluminescence Immunoassay (CLIA) System is powerful system with a throughput of 120 Tests/Hr. The Measurement Principle is based on Micron super paramagnetic particles (SPMP) platform with alkaline phosphatase (ALP) labeled reagents and AMPPD substrate.
Further, When there is single separation, the residual of labeled antibody would lead to false positive results; but we employ up to 4-phase wash separation steps, which would lower the possibility to the lowest level.