Order Entry
India
ContactUsLinkComponent
BeneSphera™, Hepatitis C Virus (HCV) Rapid Test
BeneSphera™, Hepatitis C Virus (HCV) Rapid Test
Catalog # BENP15001281
Supplier:  BENESPHERA
CAS Number:  
BeneSphera™, Hepatitis C Virus (HCV) Rapid Test
Catalog # BENP15001281
Supplier:  BENESPHERA
CAS Number:  
Specifications
  • Pk:
    30 Tests
  • Description:
    BeneSphera™ HCV Rapid Test
Specifications
About this item
BeneSphera™ Hepatitis C Virus (HCV) Rapid Test is a third generation kit for qualitative detection of antibodies against HCV in serum or plasma.