Order Entry
India
ContactUsLinkComponent
BeneSphera™, cTroponin I Rapid Test
BeneSphera™, cTroponin I Rapid Test
Catalog # BENP15001326
Supplier:  BENESPHERA
CAS Number:  
BeneSphera™, cTroponin I Rapid Test
Catalog # BENP15001326
Supplier:  BENESPHERA
CAS Number:  
Specifications
  • Pk:
    30 Tests
  • Description:
    BeneSphera™ cTroponin I Rapid Test
Specifications
About this item
BeneSphera™ cTroponin I Rapid Test is a rapid, qualitative test for the detection of cardiac troponin (cTroponin) in serum, plasma or whole blood as an aid in the diagnosis of myocardial infarction in emergency room, critical care, point of care and hospital settings. It consists of a coloured conjugate pad containing colloidal gold coupled with mouse monoclonal anti-cTnl antibodies and nitrocellulose membrane containing a test line (T line) and a control line (C line).
  • Detects cardiac marker cTroponin I
  • Analytical sensitivity: 0,5 ng/ml
No interference was experienced by Bilirubin up to 10 mg/dl, Cholesterol up to 800 mg/dl, Hemoglobin up to 250 mg/dl and Triglycerides up to 1250 mg/dl.