BeneSphera™ Dengue IgG/IgM Rapid Card Test is for the differential detection of IgG and IgM antibodies.
- Differential detection of IgG and IgM antibodies
- Detection of all serotypes (DEN - 1, DEN -2 , DEN - 3, DEN - 4)
- No cross reactivity
- Sensitivity: 95,5%
- Specificity: 96,4%
BeneSphera™ Dengue IgG/IgM Rapid Card Test is a solid phase immunochromatographic assay for the rapid, qualitative and differential IgG/lgM antibodies to dengue virus in human serum, plasma or whole blood. This test is intended for professional use as aid in the presumptive diagnosis between primary and secondary dengue infection
This test provides only a preliminary test result. Therefore, isolation of virus, antigen detection in fixed tissues, RT-PCR and serological test like hemagglutination-inhibition test, more specific alternative diagnosis method must be used in order to obtain a confirmation of dengue virus Infection. This kit can detect all 4 Dengue serotypes by using a mixture of recombinant dengue envelope proteins.