Order Entry
India
ContactUsLinkComponent
BeneSphera™ E-21, ELISA microplate reader, multi-mode
BeneSphera™ E-21, ELISA microplate reader, multi-mode
Supplier:  BENESPHERA
Certificates
undefined
BeneSphera™ E-21, ELISA microplate reader, multi-mode
Supplier:  BENESPHERA

Specifications

  • Catalog No:
  • BENP20001999
  • BENP20001753
  • Light source:
  • OSRAM 64607
  • Wavelength range:
  • 405, 450, 492, 630 nm
  • Voltage:
  • 110 V - 250 V
  • W×D×H:
  • 330×450×140 mm
  • Weight:
  • 10 kg

Specifications

Related Information

Frequently Bought Together