Order Entry
India
ContactUsLinkComponent
BeneSphera™ C-71, Clinical chemistry Analyser
  BENP20001752
BeneSphera™ C-71, Clinical chemistry Analyser
  BENP20001752
  • Description:
    Clinical chemistry analyser BeneSphera™ C-71
  • Pk:
    1
BeneSphera™ C-71 is a fully automated, discrete, random access clinical chemistry analyser.
  • Throughput: Capable of running up to 100 tests/hour and up to 300 tests/hour with ISE (Na+/K+/Li+/Cl+)
  • Up to 28 onboard chemistries
  • Flexible: 36 sample and reagent positions
  • Refrigerated reagent compartment
  • Automatic probe cleaning, liquid level detection and collision protection
  • Eight wavelengths: 340 to 670 nm
  • Automatic dilution for abnormal samples
  • Bi-directional LIS interface