Order Entry
India
Orders LinkContactUsLinkComponent
VWR® VistaVision, Microscope slides, colour-coded
VWR® VistaVision, Microscope slides, colour-coded
Supplier:   VWR Collection
VWR® VistaVision, Microscope slides, colour-coded
Supplier:  VWR Collection
Certificates
VWR® VistaVision, Microscope slides, colour-coded
2 Options Available Below
Product Details & Documents
Bright printed colours of these microscope slides provide an ideal marking area and enable easy identification. The ultra-thin colour frosted end facilitates processing on automated systems.
  • Manufactured using high quality, white soda lime glass
  • Pre-cleaned and ready for use
  • Autoclavable
  • Colour coded ends facilitate sorting and identification
  • Uniform size and thickness
 : ISO 8037/1
Product Family Options
Product Information
  • Colour
    Thickness
    L×W
    Availability
    Price
  • VWRU16004-382
    Microscopy slides, premium, square corners
    Microscopy slides, premium, square corners
    White
    1,0 mm
    75×25 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items
  • VWRU16004-384
    Microscopy slides, premium, square corners
    Microscopy slides, premium, square corners
    Blue
    1,0 mm
    75×25 mm
    Log in for availability
    Log in for availability
    1 * 1.440 items
    1 * 1.440 items