VWR®SuperFrost®, Microscope slides
VWRU48311-601
- Colour:White
- Description:Microscope slides, SuperFrost
- L×W:75×25 mm
- Seq:01
- Thickness:1 mm
- Pk:72
These microscope slides come with a light opaque coating in different colours.
- Resistant to commonly used solvents
- No scratching or sticking when stacked together
- Label areas on the top of the slide, for quick identification
Coating takes and retains most laboratory marking methods better than plain or conventional frosted slides. The coating is impervious to most common laboratory chemicals and reagents. The raised printed writing surface eliminates the need for paper labels and re-labelling of slides.