Order Entry
India
ContactUsLinkComponent
VWR®, Beta Radiation Safety Rack, 8-Well
VWR®, Beta Radiation Safety Rack, 8-Well
Catalog # VWRU82010-758
Supplier:  VWR Collection
undefined
VWR®, Beta Radiation Safety Rack, 8-Well
Catalog # VWRU82010-758
Supplier:  VWR Collection
Specifications
  • W×D×H:
    90×37×55 mm
  • Pk:
    1
Specifications
About this item
Made of 8 mm thick acrylic, this sturdy and solid rack holds eight 1,5 to 2,0 ml microcentrifuge tubes.
  • Sturdy transportation
  • Snug lid ensures secure fit
Ideal for safe transportation or benchtop storage of beta-emitting radioactive isotope samples.