VWR®, Stoppers for Chlorobutyl Serum and Lyophilisation
Supplier: VWR Collection
Certificates
About this item
These stoppers have a fluorocarbon film on the top and bottom. In addition, the enhanced lyophilisation stoppers are coated with a cross-linkable polydimethylsiloxane on the underside of the flange.
- Coating helps to reduce friction as stoppers pass through filling lines and helps reduce the level of subvisible particulate matter
- Enhanced stoppers are also ready for sterilisation without further preparation
- Stoppers are moulded from elastomeric formula 4432/50 grey
This formula has extremely low levels of organic and metallic extractables and does not contain plasticizers, 2-Mercaptobenzothiazole, nitrosamine precursors, or natural rubber latex.
Certifications: All stoppers are manufactured in cGMP facilities and tested for quality. Stoppers meet USP, Ph. Eur., and JP requirements.
Packaging: They are packaged in double sterilizable bags designed for direct entry into sterilisation units. Each shipment also includes a certificate of analysis.
Frequently Bought Together