Order Entry
India
ContactUsLinkComponent
VWR®, Cellulose Sponges, Fine-Pore
VWR®, Cellulose Sponges, Fine-Pore
Catalog # VWRU58542-360
Supplier:  VWR Collection
undefined
VWR®, Cellulose Sponges, Fine-Pore
Catalog # VWRU58542-360
Supplier:  VWR Collection
Specifications
  • Description:
    Cellulose sponges, uncompressed, expanded dimensions of 152×114×44 mm
  • Seq:
    02
  • Pk:
    6
Specifications
About this item
These fine pore natural colour cellulose sponges hold approximately 25 times their own weight in water.
  • Durable
  • Convenient for quick laboratory wipe-up
Resist tearing and abrasion. Absorb liquid more evenly than large pore sponges.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904