VWR® Signature™, Pipette Tips, Extra Length
Catalog # VWRU37001-522
Supplier: VWR Collection
Specifications
- Packed:96/hinged rack
- Sterile:-
- Volume:1 - 200 µl
- Length:84 mm
- Seq:01
- Pk:576
Specifications
About this item
Clear polypropylene, extra-long tips feature a uniquely thin and flexible top that conforms to the pipettor barrel, forming a tight seal with minimal ejection force.
- The unbeveled point is extremely precise to allow easy dispensing of the smallest sample
- Tips and racks are autoclavable
Tips are lot-certified pure and free of human DNA, RNase, DNase, endotoxins, and heavy metals.