VWR®, Media/Storage Bottles, with GL Screw Caps
Media/Storage bottles are designed with a wide opening (30 mm internal diameter) and a drip-free polypropylene pour ring.
- Very good chemical resistance
- High thermal shock resistance
- Scale graduations
- Autoclavable
These bottles are manufactured from high quality type 1, class A, 33 expansion borosilicate glass. It can be autoclaved to 180 °C (356 °F). They feature enhanced graduations and marking spots printed in permanent white colour.
: All bottles are available with or without a GL 45 polypropylene cap, with the exception of the 50 ml bottles, which feature a GL 32 thread.
Frequently Bought Together