Order Entry
India
ContactUsLinkComponent
VWR®Spinbar®, Magnetic stirring bars, with pivot ring, giant polygon
VWR®Spinbar®, Magnetic stirring bars, with pivot ring, giant polygon
Catalog # VWRU74950-280
Supplier:  VWR Collection
undefined
VWR®Spinbar®, Magnetic stirring bars, with pivot ring, giant polygon
Catalog # VWRU74950-280
Supplier:  VWR Collection

Specifications

  • Ø:
    15,9 mm
  • Type:
    With pivot ring
  • Length:
    127 mm
  • Seq:
    16
  • Pk:
    1

Specifications

About this item

Designed for stirring large volumes in vessels such as drums and tanks.

  • Marring of plastic containers is diminished
  • Multifaceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Ideal for use with Cool Stir® large volume stirrer
  • Autoclave safe

A moulded pivot ring minimises the contact area between the bar and the vessel, reducing friction and chattering.

Certifications: Alnico magnet encapsulated in FDA grade, USP class VI PTFE coating – superior chemical compatibility. Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification.