EDTA disodium salt 0.05 M (M/20) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Density:1,01 g/cm³
EINECS:205-358-3
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:—
- Molarity:0.0495 - 0.0505
Specification Test Results
Frequently Bought Together