Soda lime, BAKER ANALYZED® ACS (4-8 Mesh) indicator, J.T.Baker®
Catalog # 3447-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- ACS - Loss on Drying at 200°C:≤7 %
- ACS - Carbon Dioxide Absorption Capacity:≥19 %
- ACS - Presence of Indicator:Passes Test
- ACS - Fines:≤1 %
- Mesh - On U.S. No. 4 Sieve:≤10 %
- Mesh - Thru U.S. No. 8 Sieve:≤25 %
Specification Test Results
About this item
:
UN:1907
ADR:8,III
:GHS07
CAS Number:8006-28-8
MDL Number:MFCD00134094