Trifluoroacetic acid 0,05% (v/v) in water, BAKER ANALYZED® LC-MS, J.T.Baker®
Catalog # 9839-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:—
- Appearance:Passes Test
- Trifluoroacetic Acid:0.045 - 0.055 %
- Gradient Elution Test (a.u.) - 254 nm:≤0.01
- Residue after Evaporation:≤1.0 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤50 ppb
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - * Calcium (Ca):≤50.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - Silicon (Si):≤50.0 ppb
- Trace Impurities - * Sodium (Na):≤50.0 ppb
Specification Test Results
About this item
:
:114.02 g/mol
Density:1,00 g/cm³
:Filtered through a 0.2 µm filter.
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169