

Trichloroethylene ≥99.5% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
9458-01
: Avantor
:
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
- ACS - Assay (ClCH:CCl₂) (by GC, corrected for water):≥ 99.5 %
- ACS - Color (APHA):≤ 10
- ACS - Residue after Evaporation:≤ 5 ppm
- ACS - Free Halogens:Passes Test
- ACS - Water (H₂O)(by coulometry):≤ 0.005 %
- ACS - Titrable Acid (meq/g):≤ 0.0001
- ACS - Titrable Base (meq/g):≤ 0.0003
- Heavy Metals (as Pb):≤ 0.1 ppm
- Trace Impurities - Copper (Cu):≤ 0.10 ppm
- Trace Impurities - Nickel (Ni):≤ 0.1 ppm
:
:12,09769
:86,7 °C (1013 hPa)
:–73 °C
:1,46 g/cm³ (20 °C)
:6.1,III
:Management of Change (MOC) category = N
:131.39 g/mol
:Ambient
:201-167-4
:1710
:GHS08
:GHS07
:79-01-6
:ClCH=CCl₂
:MFCD00000838