


2-Propanol ≥99.5%, AR® ACS for histology, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:Management of Change (MOC) category = N
Flash Pt:12 °C
REACH:01-2119457558-25
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance (clear, colorless liquid):Passes Test
- Infrared Spectrum (Conforms to Standard):Passes Test
- ACS - Assay (CH₃CHOHCH₃):≥99.5 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Titrable Acid or Base (meq/g):≤0.0001
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.2 %
- Carbonyl Compounds - Propionaldehyde:≤0.002 %
- Carbonyl Compounds - Acetone:≤0.002 %
Specification Test Results
Frequently Bought Together