Order Entry
India
ContactUsLinkComponent
Dichloromethane ≥99.5% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Irritant
Health
Dichloromethane ≥99.5% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Health
Dichloromethane ≥99.5% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • ACS - Assay (CH₂Cl₂) (by GC, corrected for water):
    ≥99.5 %
  • Appearance:
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Free Halogens:
    Passes Test
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Titrable Acid (µeq/g):
    ≤0.2
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340-400 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.35
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm:
    ≤1.00
  • Water (by KF, coulometric):
    ≤0.02 %

Specification Test Results

Related Information

Frequently Bought Together