


Hexachloroplatinic(IV) acid solution 10%, BAKER ANALYZED®, J.T.Baker®
Catalog # 2896-00
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:30 g
- Seq:3332
Specifications
Specification Test Results
- Assay (as Pt):3.77 - 3.87 %
Specification Test Results
About this item
:
:409.81 g/mol
Density:1,00 g/cm³
UN:3264
ADR:8,II
:GHS05
:GHS07
:GHS08
CAS Number:16941-12-1
Formula:H₂Cl₆Pt
MDL Number:MFCD00011330