Potassium nitrite ≥96.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
3202-01
: Avantor
:
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:500 g
- Seq:3645
- Assay (KNO₂) (by KMnO₄ titrn):≥96.0 %
- Insoluble Matter:≤0.01 %
- pH of 5% Solution at 25°C:7.0 - 10.0
- Chloride (Cl):≤0.02 %
- Sulfate (SO₄):≤0.01 %
- Calcium (Ca):≤0.005 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Sodium (Na):≤0.5 %
- ACS - Magnesium (Mg):≤0.002 %
:
:85.1 g/mol
:13,07734
:387 °C
:1,92 g/cm³ (20 °C)
:Ambient
:231-832-4
:1488
:5.1,II
:GHS03
:GHS06
:GHS09
:7758-09-0
:KNO₂
:MFCD00011408