J.T.Baker®, Bovine Serum Albumin (BSA), Fraction V
Catalog # A464-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Wide mouth amber glass bottle, black tamper evident cap
- Pk:10 g
- CAS No.:9048-46-8
- Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Colour:light yellow