J.T.Baker® BAKER INSTRA-ANALYZED®, Palladium, 1000 µg/ml (0.1% w/v), AA Standard
Catalog # 6462-04
Supplier: Avantor
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pack type:PE bottle
- Pk:150 mL
- Accuracy:±0,3%.
- Element:Palladium
- Symbol:Pd
Specifications
About this item
Palladium standard is prepared by dissolution of palladium metal of 99,95% spectral purity.The metal content of the solution is confirmed by ICP analysis, traceable to NIST SRM 3138.
- Standardised with NIST standard chemicals