Order Entry
India
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Mercury AA Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Mercury AA Standard
Catalog # 6459-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Mercury AA Standard
Catalog # 6459-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly bottle for liquids, narrow mouth
  • Pk:
    150 mL
  • Accuracy:
    ±0.3%.
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Mercury standard prepared by dissolution of mercury metal of 99,998% spectral purity.The metal content of the solution is confirmed by titration with EDTA,traceable to NIST SRM 915.

  • Standardised with NIST standard chemicals