Order Entry
India
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Chromium AA Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Chromium AA Standard
Catalog # 6449-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Chromium AA Standard
Catalog # 6449-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Poly bottle for liquids, narrow mouth
  • Pk:
    150 mL
  • Element:
    Chromium
  • Symbol:
    Cr

Specifications

About this item

Prepared by dissolution of chromium metal of 99,99% spectral purity. The metal content of the solution isconfirmed by ICP analysis, traceableto NIST SRM 3112a.

  • Standardised with NIST standard chemicals